missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Yellow Color Code Counter Shutters

Designed for use with Eppendorf Pipettes

Brand:  Eppendorf™ 3121810009

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 15244386

  • 43.00 SEK / Each


Estimated Shipment
06-09-2018
Add to basket

Description

Description

  • Accessory for Eppendorf pipettes
  • For use with liquid handling applications
  • Counter shutters available for fixed and variable pipettes
Specifications

Specifications

For 200μL Single-channel Fixed Pipette
1Each
Color Code Counter Shutter
Videos