Learn More
Clark Scientific™ PVC Yellow Spill Trays
Manufactured from rigid PVC, these small yellow spill trays are for use in radiation areas.
Brand: Clark Scientific™ YPVC Small
Additional Details : Weight : 0.55084kg
Description
- Available in yellow for use in radiation areas
- Avaialble in white for use outside radiation areas
- Colorless PVC liners available separately
- Manufactured from rigid PVC
Order Info
Size
Specifications
Yellow | |
Spill Tray Bench | |
1 Ea. |
PVC | |
Radiation areas | |
Small |