Läs mer
Tocris Bioscience™ Yoda 1
Piezo1 channel activator
Brand: Tocris Bioscience™ 5586/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Yoda 1 is a piezo1 channel activator. Slows the inactivation phase of transient currents, sensitizes Piezo1 to activation by pressure, and partially activates channels in the absence of external pressure, in HEK cells expressing Piezo1 channels.Specifikationer
| 2-[5-[[(2,6-Dichlorophenyl)methyl]thio]-1,3,4-thiadiazol-2-yl]pyrazine | |
| 448947-81-7 | |
| 50 mg |