missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YTHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14392-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Function: May be part of a signal transduction pathway that influences splice site selection.Specifications
YTHDC1 | |
Polyclonal | |
Unconjugated | |
YTHDC1 | |
KIAA1966, Putative splicing factor YT521, YT521, YT521 B, YTH domain containing 1, YTHDC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170956, 231386, 91746 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
E9Q5K9, Q96MU7, Q9QY02 | |
Ythdc1 | |
YTHDC1 Fusion Protein Ag5882 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |