missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049008
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]

Used for phosphors, ceramic capacitors, ferrite devices, pulsed lasers and catalysts. Use in improving the strength and mechanical properties of stainless steel. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
UN Number UN3178
Quantity 25 x 25 mm
Solubility Information It reacts slowly with water and is soluble in dilute acids.
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.43g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Ytterbium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'