Läs mer
Thermo Scientific™ Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Kapacitet (metrisk):
50 ml
100 ml
200 ml
Diameter (metrisk) Yttre:
51 mm
62 mm
77 mm
Höjd (metrisk):
58 mm
72 mm
90 mm
Förpackningsstorlek:
Varje
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| Material | Zirconium Oxide |
| Kvantitet | 1 Piece |
| Kapacitet (metrisk) | 50 ml |
| Diameter (metrisk) Yttre | 51 mm |
| Höjd (metrisk) | 58 mm |
| Produkttyp | Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form |
| Diameter (metrisk) bas | 34mm |