Läs mer
YU238259, MedChemExpress
Produktkod. 16493917
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493917 50 mg
16460458 5 mg
16437776 100 mg
16427998 10 mg
16413726 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16493917 Leverantör Medchem Express Leverantörsnummer HY19977
MedChemExpress YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research.
YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research.
Specifikationer
Kemiskt namn eller material YU238259
CAS 1943733-16-1
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H22ClN3O4S
Kvantitet 50 mg
Löslighetsinformation DMSO : 110 mg/mL (239.16 mM; Need ultrasonic)
LEDER COC1=CC=C(S(NCC2=CC=C(C(NCCC3=CC=C(Cl)C=N3)=O)C=C2)(=O)=O)C=C1
Molekylvikt (g/mol) 459.95
Formel vikt 459.95
Procent renhet 99.57%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease