Läs mer
(Z)-10-Tritriacontene, TRC
Produktkod. 30454058
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454058 100 mg
30456601 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT456820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-10-Tritriacontene
CAS 99026-87-6
Molekylformel C33 H66
Kvantitet 100 mg
Synonym (10Z)-10-Tritriacontene
InChI formel InChI=1S/C33H66/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h19,21H,3-18,20,22-33H2,1-2H3/b21-19-
LEDER CCCCCCCCCCCCCCCCCCCCCC\C=C/CCCCCCCCC
IUPAC-namn (Z)-tritriacont-10-ene
Molekylvikt (g/mol) 462.877
Formel vikt 462.516