Läs mer
(Z)-11-Hexadecenoic Acid, TRC
Produktkod. 30137024
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137024 25 mg
30136947 10 mg
30010565 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137024 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29396825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2416-20-8
Molekylformel C16 H30 O2
Molekylvikt (g/mol) 254.41
Synonym (11Z)-11-Hexadecenoic Acid, Hexadec-11(Z)-enoic Acid, cis-11-Hexadecenoic Acid, δ11-Hexadecenoic Acid
IUPAC-namn (Z)-hexadec-11-enoic acid
LEDER CCCC\C=C/CCCCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material (Z)-11- Hexadecenoic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h5-6H,2-4,7-15H2,1H3,(H,17,18)/b6-5-
Formel vikt 254.2246