Läs mer
(Z)-13-Octadecen-1-ol, TRC
Produktkod. 30178954
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30178954 25 mg
30179671 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30178954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23554025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69820-27-5
Molekylformel C18 H36 O
Molekylvikt (g/mol) 268.48
Synonym (13Z)-Octadecen-1-ol, cis-13-Octadecenol;
IUPAC-namn (Z)-octadec-13-en-1-ol
LEDER CCCC\C=C/CCCCCCCCCCCCO
Specifikationer
Kemiskt namn eller material (Z)-13-Octadecen-1-ol
Kvantitet 25 mg
InChI formel InChI=1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h5-6,19H,2-4,7-18H2,1H3/b6-5-
Formel vikt 268.2766