Läs mer
(Z)-13-Octadecen-1-ol, TRC
Produktkod. 30179671
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179671 250 mg
30178954 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179671 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO235540250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69820-27-5
Molekylformel C18 H36 O
Molekylvikt (g/mol) 268.48
Synonym (13Z)-Octadecen-1-ol, cis-13-Octadecenol;
IUPAC-namn (Z)-octadec-13-en-1-ol
LEDER CCCC\C=C/CCCCCCCCCCCCO
Specifikationer
Kemiskt namn eller material (Z)-13-Octadecen-1-ol
Kvantitet 250 mg
InChI formel InChI=1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h5-6,19H,2-4,7-18H2,1H3/b6-5-
Formel vikt 268.2766