Läs mer
(Z)-2-Chloropropane-1,3-diyl Dioleate, TRC
Produktkod. 17972435
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972435 25 mg
17991785 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17972435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38030525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1639207-42-3
Molekylformel C39 H71 Cl O4
Molekylvikt (g/mol) 639.43
IUPAC-namn [2-chloro-3-[(Z)-octadec-9-enoyl]oxy-propyl] (Z)-octadec-9-enoate
LEDER CCCCCCCC\C=C/CCCCCCCC(=O)OCC(Cl)COC(=O)CCCCCCC\C=C/CCCCCCCC
Specifikationer
Kemiskt namn eller material (Z)-2-Chloropropane-1,3-diyl Dioleate
Kvantitet 25 mg
InChI formel InChI=1S/C39H71ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37H,3-16,21-36H2,1-2H3/b19-17-,20-18-
Formel vikt 638.5041