Läs mer
(Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, TRC
Produktkod. 30452165
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30452165 500 mg
1 options
Produktkod. 30452165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ701733500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-(Furan-2-yl)(methoxyimino)acetic Acid
CAS 39684-61-2
Molekylformel C7 H7 N O4
Kvantitet 500 mg
Synonym (Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, 2-Furanacetic acid','α-(methoxyimino)-','(αZ)-, 2-Furanacetic acid','α-(methoxyimino)-','(Z)-, (αZ)-α-(Methoxyimino)-2-furanacetic acid, (Z)-2-(2-Furyl)-2-methoxyiminoacetic acid, syn-2-Furyl-2-methoxyiminoacetic acid, (2Z)-2-(2-Furyl)-2-methoxyiminoacetic Acid, (Z)-2-(Furan-2-yl)-2-(methoxyimino)acetic Acid, (Z)-(Furan-2-yl)(methoxyimino)acetic Acid
InChI formel InChI=1S/C7H7NO4/c1-11-8-6(7(9)10)5-3-2-4-12-5/h2-4H,1H3,(H,9,10)/b8-6-
LEDER CO\N=C(/C(=O)O)\c1occc1
IUPAC-namn (2Z)-2-(furan-2-yl)-2-methoxyiminoacetic acid
Molekylvikt (g/mol) 169.135
Formel vikt 169.038