Läs mer
(Z)-2-(Methoxyimino)-3-oxo-butanoic Acid Ethyl Ester, TRC
Produktkod. 30475462
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475462 100 mg
1 options
Produktkod. 30475462 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM313255100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-2-(Methoxyimino)-3-oxo-butanoic Acid Ethyl Ester
CAS 66340-86-1
Molekylformel C7H11NO4
Kvantitet 100 mg
Synonym (2Z)-2-(Methoxyimino)-3-oxo-butanoic Acid Ethyl Ester, Ethyl syn-2-Methoxyiminoacetoacetate
InChI formel InChI=1S/C7H11NO4/c1-4-12-7(10)6(5(2)9)8-11-3/h4H2,1-3H3/b8-6-
LEDER CC(/C(C(OCC)=O)=N/OC)=O
IUPAC-namn ethyl (2Z)-2-methoxyimino-3-oxobutanoate
Molekylvikt (g/mol) 173.17
Formel vikt 173.0688