Läs mer
(Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine Maleate, Mikromol™
Produktkod. 30049891
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30049891 50 mg
1 options
Produktkod. 30049891 Leverantör Mikromol Leverantörsnummer MM0032.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine Maleate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine Maleate
CAS 147365-12-6
Analyt- eller komponentnamn (Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine Maleate
Frakt skick Room Temperature
Molekylformel C19 H21 N O . C4 H4 O4
Certifikat och efterlevnad ISO 9001
Synonym Doxepine Imp. D (EP) as Maleate, (Z)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N,N-dimethylpropan-1-amine Maleate, 1-Propanamine, 3-dibenz[b,e]oxepin-11(6H)-ylidene-N,N-dimethyl-, (Z)-, (Z)-2-butenedioate, Dibenz[b,e]oxepin, 1-propanamine deriv, Doxepin Hydrochloride Imp. D (EP), Doxepin Imp. D (EP), Doxepin Hydrochloride Impurity D as Maleate, Doxepin Impurity D as Maleate
InChI formel InChI=1S/C19H21NO.C4H4O4/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;5-3(6)1-2-4(7)8/h3-6,8-12H,7,13-14H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b17-11-;2-1-
LEDER CN(C)CC\C=C/1\c2ccccc2COc3ccccc13.OC(=O)\C=C/C(=O)O
IUPAC-namn (3Z)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;(Z)-but-2-enedioic acid
Molekylvikt (g/mol) 395.45
Kvantitet 50 mg
Formel vikt 395.1733 g/mol