Läs mer
(Z)-4-Hydroxy Tamoxifen, TRC
Produktkod. 30134553
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134553 100 mg
30004247 5 mg
30011214 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134553 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH954725100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68047-06-3
Molekylformel C26H29NO2
Molekylvikt (g/mol) 387.51
Synonym Tamoxifen-4-hydroxy, 4-[(1Z)-1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]phenol (ACI), Phenol, 4-[(1Z)-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]- (9CI), Phenol, 4-[1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]-,
IUPAC-namn 4-[(Z)-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenylbut-1-enyl]phenol
LEDER CCC(=C(c1ccc(O)cc1)c2ccc(OCCN(C)C)cc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Z-4-Hydroxy Tamoxifen
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C26H29NO2/c1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(28)15-11-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25-
Formel vikt 387.2198
Procent renhet >95%