Läs mer
Z-ALA-ALA-NH2, TRC
Produktkod. 30328006
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328006 100 mg
1 options
Produktkod. 30328006 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA576043100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Z-ALA-ALA-NH2
CAS 50444-54-7
Molekylformel C14H19N3O4
Kvantitet 100 mg
Synonym N-[(Phenylmethoxy)carbonyl]-L-alanyl-L-alaninamide, N-Benzyloxycarbonyl-L-alanyl-L-alaninamide, N-Carbobenzoxyalanylalaninamide, NSC 164062
InChI formel InChI=1S/C14H19N3O4/c1-9(12(15)18)16-13(19)10(2)17-14(20)21-8-11-6-4-3-5-7-11/h3-7,9-10H,8H2,1-2H3,(H2,15,18)(H,16,19)(H,17,20)/t9-,10-/m0/s1
LEDER C[C@H](NC(=O)[C@H](C)NC(=O)OCc1ccccc1)C(=O)N
IUPAC-namn benzyl N-[(2S)-1-[[(2S)-1-amino-1-oxopropan-2-yl]amino]-1-oxopropan-2-yl]carbamate
Molekylvikt (g/mol) 293.32
Formel vikt 293.1376