Läs mer
Z-ALA-ALA-NH2, TRC
Specifications
Specifications
| Kemiskt namn eller material | Z-ALA-ALA-NH2 |
| CAS | 50444-54-7 |
| Molekylformel | C14H19N3O4 |
| Kvantitet | 500 mg |
| Synonym | N-[(Phenylmethoxy)carbonyl]-L-alanyl-L-alaninamide, N-Benzyloxycarbonyl-L-alanyl-L-alaninamide, N-Carbobenzoxyalanylalaninamide, NSC 164062 |
| InChI formel | InChI=1S/C14H19N3O4/c1-9(12(15)18)16-13(19)10(2)17-14(20)21-8-11-6-4-3-5-7-11/h3-7,9-10H,8H2,1-2H3,(H2,15,18)(H,16,19)(H,17,20)/t9-,10-/m0/s1 |
| LEDER | C[C@H](NC(=O)[C@H](C)NC(=O)OCc1ccccc1)C(=O)N |
| IUPAC-namn | benzyl N-[(2S)-1-[[(2S)-1-amino-1-oxopropan-2-yl]amino]-1-oxopropan-2-yl]carbamate |
| Molekylvikt (g/mol) | 293.32 |
| Formel vikt | 293.1376 |