Läs mer
Z-Cys(Bzl)-OH, TRC
Produktkod. 30356191
Change view
Kvantitet:
500 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30356191 500 mg -
30360393 2.5 g 2.5g
30358218 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356191 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC994800500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3257-18-9
Molekylformel C18H19NO4S
Molekylvikt (g/mol) 345.41
IUPAC-namn (2R)-3-benzylsulfanyl-2-(phenylmethoxycarbonylamino)propanoic acid
LEDER OC(=O)[C@H](CSCc1ccccc1)NC(=O)OCc2ccccc2
Specifikationer
Kemiskt namn eller material Z-Cys(Bzl)-OH
InChI formel InChI=1S/C18H19NO4S/c20-17(21)16(13-24-12-15-9-5-2-6-10-15)19-18(22)23-11-14-7-3-1-4-8-14/h1-10,16H,11-13H2,(H,19,22)(H,20,21)/t16-/m0/s1
Kvantitet 500 mg
Formel vikt 345.1035