Läs mer
(Z)-Fluoxastrobin, TRC
Produktkod. 30137345
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137345 1 mg
30134609 10 mg
17991035 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF5967011MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 887973-21-9
Molekylformel C21 H16 Cl F N4 O5
Molekylvikt (g/mol) 458.83
Synonym Methanone, [2-[[6-(2-chlorophenoxy)-5-fluoro-4-pyrimidinyl]oxy]phenyl](5,6-dihydro-1,4,2-dioxazin-3-yl)-, O-methyloxime, (1Z)-, (Z)-Fluoxastrobin
IUPAC-namn (Z)-1-[2-[6-(2-chlorophenoxy)-5-fluoropyrimidin-4-yl]oxyphenyl]-1-(5,6-dihydro-1,4,2-dioxazin-3-yl)-N-methoxymethanimine
LEDER CO\N=C(/C1=NOCCO1)\c2ccccc2Oc3ncnc(Oc4ccccc4Cl)c3F
Specifikationer
Kemiskt namn eller material (Z)-fluoxastrobin
Kvantitet 1 mg
InChI formel InChI=1S/C21H16ClFN4O5/c1-28-26-18(21-27-30-11-10-29-21)13-6-2-4-8-15(13)31-19-17(23)20(25-12-24-19)32-16-9-5-3-7-14(16)22/h2-9,12H,10-11H2,1H3/b26-18-
Formel vikt 458.0793