missing translation for 'onlineSavingsMsg'
Läs mer

(Z)-Hexadec-11-en-1-ol, TRC

Produktkod. 30180364
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180364 100 mg
30179979 1 g
30179853 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30180364 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH291114100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 56683-54-6
Molekylformel C16 H32 O
Molekylvikt (g/mol) 240.42
Synonym 11-Hexadecen-1-ol, (Z)-, (11Z)-11-Hexadecen-1-ol, (Z)-11-Hexadecen-1-ol, (Z)-11-Hexadecenol, cis-11-Hexadecen-1-ol
IUPAC-namn (Z)-hexadec-11-en-1-ol
LEDER CCCC\C=C/CCCCCCCCCCO

Specifikationer

Kemiskt namn eller material (Z)-11-Hexadecen-1-ol
Kvantitet 100 mg
InChI formel InChI=1S/C16H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h5-6,17H,2-4,7-16H2,1H3/b6-5-
Formel vikt 240.2453
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.