Läs mer
(Z)-Methyl 2-(3-Nitrobenzylidene)-3-oxobutanoate, TRC
Produktkod. 30465947
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30465947 5 g
1 options
Produktkod. 30465947 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3035805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-Methyl 2-(3-Nitrobenzylidene)-3-oxobutanoate
CAS 119128-13-1
Molekylformel C12H11NO5
Kvantitet 5 g
Synonym (Z)-2-[(3-Nitrophenyl)methylene]-3-oxobutanoic Acid Methyl Ester, (2Z)-2-[(3-Nitrophenyl)methylene]-3-oxobutanoic Acid Methyl Ester
InChI formel InChI=1S/C12H11NO5/c1-8(14)11(12(15)18-2)7-9-4-3-5-10(6-9)13(16)17/h3-7H,1-2H3
LEDER COC(=O)\C(=C/c1cccc(c1)[N+](=O)[O-])\C(=O)C
Molekylvikt (g/mol) 249.22