Läs mer
(Z)-Naftifine, TRC
Produktkod. 30132827
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132827 50 mg
30011714 100 mg
30007730 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132827 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN21310550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65473-08-7
Molekylformel C21H21N
Molekylvikt (g/mol) 287.4
Synonym (Z)-N-Methyl-N-(3-phenyl-2-propen-1-yl)-1-naphthalenemethanamine Hydrochloride, (Z)-N-Cinnamyl-N-methyl-1-naphthalenemethanamine Hydrochloride, (Z)-AW 105-843, (Z)-Exoderil, (Z)-Naftifungin, (Z)-Naftin, (Z)-SN 105-843
IUPAC-namn (Z)-N-methyl-N-(1-naphthylmethyl)-3-phenyl-prop-2-en-1-amine
LEDER CN(C\C=C/c1ccccc1)Cc2cccc3ccccc23
Specifikationer
Kemiskt namn eller material (Z)-Naftifine
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C21H21N/c1-22(16-8-11-18-9-3-2-4-10-18)17-20-14-7-13-19-12-5-6-15-21(19)20/h2-15H,16-17H2,1H3/b11-8-
Formel vikt 287.1674
Procent renhet >95%