Läs mer
(Z)-Tetrachlorvinphos, TRC
Produktkod. 30413678
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413678 5 mg
30422346 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413678 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2923055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22248-79-9
Molekylformel C10 H9 Cl4 O4 P
Molekylvikt (g/mol) 363.8992561
Synonym Phosphoric acid, (1Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester, Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)ethenyl dimethyl ester, (Z)-, Phosphoric acid, 2-chloro-1-(2,4,5-trichlorophenyl)vinyl dimethyl ester, (Z)- (8CI), (Z)-Tetrachlorovinphos, CVMP, Debantic, Gardicide, Gardona, Rabon, Rabond, SD 8447, Stirofos, Stirophos, Stitifos, Tetrachlorvinfos, Tetrachlorvinphos, Vinfos, cis-Gardona
IUPAC-namn [(Z)-2-chloro-1-(2,4,5-trichlorophenyl)ethenyl] dimethyl phosphate
LEDER COP(=O)(OC)O\C(=C/Cl)\c1cc(Cl)c(Cl)cc1Cl
Specifikationer
Kemiskt namn eller material Tetrachlorvinphos
Kvantitet 5 mg
InChI formel InChI=1S/C10H9Cl4O4P/c1-16-19(15,17-2)18-10(5-11)6-3-8(13)9(14)4-7(6)12/h3-5H,1-2H3/b10-5-
Formel vikt 363.8993