Learn More
(Z)-Triprolidine Dihydrochloride, TRC
Product Code. 30456855
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456855 1 mg 1mg
30451732 5 mg 5mg
30450661 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30456855 Supplier Toronto Research Chemicals Supplier No. TRCT8140251MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (Z)-Triprolidine Dihydrochloride
Analyte or Component Names (Z)-Triprolidine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C19H22N2 • 2(HCl)
Synonym 2-[(1Z)-1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Dihydrochloride, (Z)-2-[1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Dihydrochloride
InChI Formula InChI=1S/C19H22N2.2ClH/c1-16-7-9-17(10-8-16)18(19-6-2-3-12-20-19)11-15-21-13-4-5-14-21;;/h2-3,6-12H,4-5,13-15H2,1H3;2*1H/b18-11-;;
Solution Type Neat
SMILES CC1=CC=C(/C(C2=NC=CC=C2)=C/CN3CCCC3)C=C1.Cl.Cl
IUPAC Name (Z)-2-(3-(pyrrolidin-1-yl)-1-(p-tolyl)prop-1-en-1-yl)pyridine dihydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 351.31
Quantity 1 mg
Formula Weight 350.14