Läs mer
(Z)-Triprolidine Oxalate Salt, TRC
Produktkod. 30455134
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30455134 5 mg
1 options
Produktkod. 30455134 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8140075MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (Z)-Triprolidine Oxalate Salt
CAS 14343-87-4
Molekylformel C19H22N2 . C2H2O4
Kvantitet 5 mg
Synonym 2-[(1Z)-1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Oxalate Salt, (Z)-2-[1-(4-Methylphenyl)-3-(1-pyrrolidinyl)-1-propenyl]pyridine Oxalate Salt
InChI formel InChI=1S/C19H22N2.C2H2O4/c1-16-7-9-17(10-8-16)18(19-6-2-3-12-20-19)11-15-21-13-4-5-14-21;3-1(4)2(5)6/h2-3,6-12H,4-5,13-15H2,1H3;(H,3,4)(H,5,6)/b18-11-;
LEDER Cc1ccc(cc1)\C(=C\CN2CCCC2)\c3ccccn3.OC(=O)C(=O)O
IUPAC-namn 2-[(Z)-1-(4-methylphenyl)-3-pyrrolidin-1-ylprop-1-enyl]pyridine;oxalic acid
Molekylvikt (g/mol) 368.4
Formel vikt 368.1736