Läs mer
Z-VAD-FMK, TRC
Produktkod. 30454432
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30454432 10 mg
1 options
Produktkod. 30454432 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ35987010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Z-VAD-FMK
CAS 187389-52-2
Molekylformel C22 H30 F N3 O7
Kvantitet 10 mg
Synonym 13: PN: WO2005039629 PAGE: 18 claimed protein, 3: PN: WO0158526 SEQID: 3 claimed sequence, 3: PN: WO2011028611 SEQID: 3 unclaimed protein, Z-VAD(OMe)-FMK, Z-Val-Ala-Asp(OMe)-FMK, N-[(Phenylmethoxy)carbonyl]-L-valyl-N-[(1S)-3-fluoro-1-(2-methoxy-2-oxoethyl)-2-oxopropyl]-L-alaninamide, (5S,8S,11S)-Methyl 11-(2-Fluoroacetyl)-5-isopropyl-8-methyl-3,6,9-trioxo-1-phenyl-2-oxa-4,7,10-triazatridecan-13-oate
InChI formel InChI=1S/C22H30FN3O7/c1-13(2)19(26-22(31)33-12-15-8-6-5-7-9-15)21(30)24-14(3)20(29)25-16(17(27)11-23)10-18(28)32-4/h5-9,13-14,16,19H,10-12H2,1-4H3,(H,24,30)(H,25,29)(H,26,31)/t14-,16-,19-/m0/s1
LEDER NCCC[N+](C)(C)CCC[n+]1ccc(C)cc1
Molekylvikt (g/mol) 467.49