Läs mer
(Z,Z)-5-(2-Methyl-3-phenyl-propenylidene)-4-oxo-2-thioxo-3-thiazolidineacetic Acid, TRC
Produktkod. 30481966
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481966 10 mg
30472490 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481966 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32680510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124782-63-4
Molekylformel C15H13NO3S2
Molekylvikt (g/mol) 319.4
Synonym (5Z)-5-[(2Z)-2-Methyl-3-phenyl-2-propen-1-ylidene]-4-oxo-2-thioxo-3-Thiazolidineacetic Acid, (5Z)- 5-[(2Z)-2-Methyl-3-phenyl-2-propenylidene]-4-oxo-2-thioxo-3-thiazolidineacetic Acid
LEDER S=C(S/C1=C\C(C)=C/C2=CC=CC=C2)N(CC(O)=O)C1=O
Specifikationer
Kemiskt namn eller material (Z,Z)-5-(2-Methyl-3-phenyl-propenylidene)-4-oxo-2-thioxo-3-thiazolidineacetic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C15H13NO3S2/c1-10(7-11-5-3-2-4-6-11)8-12-14(19)16(9-13(17)18)15(20)21-12/h2-8H,9H2,1H3,(H,17,18)/b10-7-,12-8-