Läs mer
Zaleplon Formamide, TRC
Produktkod. 30454467
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454467 50 mg
30456276 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ14501050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1227694-78-1
Molekylformel C16H13N5O
Molekylvikt (g/mol) 291.31
Synonym N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethyl-formamide, Zaleplon Impurity
IUPAC-namn N-[3-(3-cyanopyrazolo[1,5-a]pyrimidin-7-yl)phenyl]-N-ethylformamide
LEDER CCN(C=O)c1cccc(c1)c2ccnc3c(cnn23)C#N
Specifikationer
Kemiskt namn eller material Zaleplon Formamide
Kvantitet 50 mg
InChI formel InChI=1S/C16H13N5O/c1-2-20(11-22)14-5-3-4-12(8-14)15-6-7-18-16-13(9-17)10-19-21(15)16/h3-8,10-11H,2H2,1H3
Formel vikt 291.112