Läs mer
Zanamivir Azide Methyl Ester, TRC
Produktkod. 30449231
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30449231 10 mg
1 options
Produktkod. 30449231 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ14803010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zanamivir Azide Methyl Ester
CAS 152178-79-5
Molekylformel C12H18N4O7
Kvantitet 10 mg
Synonym 5-(Acetylamino)-2,6-anhydro-4-azido-3,4,5-trideoxy-D-glycero-D-galacto-non-2-enonic Acid Methyl Ester
InChI formel InChI=1S/C12H18N4O7/c1-5(18)14-9-6(15-16-13)3-8(12(21)22-2)23-11(9)10(20)7(19)4-17/h3,6-7,9-11,17,19-20H,4H2,1-2H3,(H,14,18)/t6-,7-,9-,10-,11-/m1/s1
LEDER CC(=O)N[C@@H]1[C@@H](C=C(O[C@H]1[C@@H]([C@@H](CO)O)O)C(=O)OC)N=[N+]=[N-]
Molekylvikt (g/mol) 330.29
Formel vikt 330.1175