missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZAP-70, His Tag Recombinant Protein
ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins.
8060.00 SEK - 18240.00 SEK
Specifications
Accession Number | NP_001070 |
---|---|
Gene Alias | ZAP-70 |
KinaseFamily | SYK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11509256
|
Invitrogen™
P2782 |
20μg |
8060.00 SEK
20µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418383
|
Invitrogen™
PR3017C |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation.
Specifications
NP_001070 | |
SYK Kinase Family | |
Recombinant | |
Baculovirus |
ZAP-70 | |
Human | |
His Tag |