Learn More
Labcon ZAP™ SLIK 100 μL Low Retention Aerosol Filter Pipette Tips
Fits Most Popular 100 μL Pipettors
Brand: Labcon 1174-965-006-9
1092.71 SEK valid until 2024-05-31
Use promo code "21712" to get your promotional price.
Additional Details : Weight : 14.96850kg
Description
- Pipette tip
- Fits Most Popular 100 μL Pipettors;
- Patented robotically inserted endotoxin free filter
- Ultra low fluid retention
- Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Made in USA
Specifications
100 μL | |
Sterile | |
Purple | |
1.95 in. | |
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Liquid Handling | |
Beveled | |
96 tips / rack | |
Yes | |
Universal Fit | |
ZAP |
Filtered | |
Natural | |
Not Autoclavable | |
49.7 mm | |
Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) | |
E-beam | |
96 tips/rack; 10 racks/Pk.; 10 Pk./case | |
RT | |
Single-use | |
No | |
Polypropylene Resin |