Läs mer
Zatebradine hydrochloride, MedChemExpress
Produktkod. 16488367
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16488367 5 mg 5mg
16445678 10 mg 10mg
16411847 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16488367 Leverantör Medchem Express Leverantörsnummer HY13422
MedChemExpress Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 μM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 μM, 2.21 μM, 1.90 μM and 1.88 μM, respectively.
Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 μM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 μM, 2.21 μM, 1.90 μM and 1.88 μM, respectively.
Specifikationer
Kemiskt namn eller material Zatebradine hydrochloride
CAS 91940-87-3
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C26H37ClN2O5
Kvantitet 5 mg
Synonym UL-FS-49 UL-FS-49CL
Löslighetsinformation DMSO : 100 mg/mL (202.82 mM; Need ultrasonic) ∣H2O : 11.11 mg/mL (22.53 mM; Need ultrasonic)
LEDER O=C1N(CCCN(CCC2=CC=C(OC)C(OC)=C2)C)CCC3=CC(OC)=C(OC)C=C3C1.Cl
Molekylvikt (g/mol) 493.04
Formel vikt 493.04
Procent renhet 98.03%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation