Läs mer
Zaurategrast ethyl ester, MedChemExpress
Produktkod. 16483766
Encompass_Preferred
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16483766 25 mg
16458198 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16483766 Leverantör Medchem Express Leverantörsnummer HY75385
MedChemExpress Zaurategrast ethyl ester (CDP323), the ethyl ester prodrug of CT7758, is a α4β1/α4β7 integrin antagonist used for the treatment of inflammatory and autoimmune disorders.
Zaurategrast ethyl ester (CDP323), the ethyl ester prodrug of CT7758, is a α4β1/α4β7 integrin antagonist used for the treatment of inflammatory and autoimmune disorders.
Specifikationer
Kemiskt namn eller material Zaurategrast ethyl ester
CAS 455264-30-9
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C28H29BrN4O3
Kvantitet 25 mg
Synonym CDP323 UCB1184197
Löslighetsinformation DMSO : 150 mg/mL (273.00 mM; Need ultrasonic)
LEDER O=C(OCC)[C@H](CC1=CC=C(NC2=NC=CC3=C2C=NC=C3)C=C1)NC4=C(Br)C(C45CCCCC5)=O
Molekylvikt (g/mol) 549.46
Formel vikt 549.46
Procent renhet 99.06%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation