missing translation for 'onlineSavingsMsg'
Learn More

ZBTB24 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27975-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825935

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May be involved in BMP2-induced transcription.
Specifications

Specifications

ZBTB24
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43167
BIF1, KIAA0441, PATZ2, ZBTB24, Zinc finger protein 450, ZNF450
Rabbit
IgG
20 μL
-20°C
Primary
9841
Western Blot
Unconjugated
ZBTB24
Liquid
ZBTB24
ZBTB24 Fusion Protein Ag24458
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers