missing translation for 'onlineSavingsMsg'
Learn More

ZDHHC7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25985-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824515

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ZDHHC7
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NXF8
DHHC 7, Palmitoyltransferase ZDHHC7, ZDHHC7, Zinc finger protein 370, ZNF370
Rabbit
IgG
20 μL
-20°C
Primary
55625
Western Blot
Unconjugated
ZDHHC7
Liquid
ZDHHC7
ZDHHC7 Fusion Protein Ag23185
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers