Läs mer
ZDHHC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24046-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.Specifikationer
| ZDHHC9 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC9 | |
| CGI 89, CXorf11, DHHC 9, DHHC9, MMSA 1, Palmitoyltransferase ZDHHC9, UNQ261/PRO298, ZDHHC10, ZDHHC9, Zinc finger protein 379, ZNF379, ZNF380 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 208884, 51114 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59268, Q9Y397 | |
| ZDHHC9 | |
| ZDHHC9 Fusion Protein Ag20881 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |