Läs mer
Zearalenone 4-Sulfate Ammonium Salt, TRC
Produktkod. 30454300
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30454300 25 mg -
30451560 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ27053025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1439328-85-4
Molekylformel C18H25NO8S
Molekylvikt (g/mol) 415.46
LEDER O=C(C1=C(O)C=C(OS(=O)([O-])=O)C=C1/C=C/CCC2)O[C@@H](C)CCCC2=O.[NH4+]
Specifikationer
Kemiskt namn eller material Zearalenone 4-Sulfate Ammonium Salt
Kvantitet 25 mg
InChI formel InChI=1S/C18H22O8S.H3N/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(26-27(22,23)24)11-16(20)17(13)18(21)25-12;/h3,7,10-12,20H,2,4-6,8-9H2,1H3,(H,22,23,24);1H3/b7-3+;/t12-;/m0./s1