Läs mer
Invitrogen™ Zenon™ Mus IgG 2a- märkningssatser
Produktkod. 10257012
Kvantitet:
25 reaktionssats
50 Reactions
50 reaktioner
50 reaktionssats
Etikett eller färgämne:
APC (Allophycocyanin)
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 594
Alexa Fluor 647
Biotin-XX
Pacific Blue
R-PE (R-Phykoerytrin)
Förpackningsstorlek:
Varje
Excitation/Emission:
410/455 nm
495/519 nm
496, 546, 565/578 nm
555/565 nm
590/617 nm
650/660 nm
650/668 nm
Produktkod. 10257012
Brand: Invitrogen™ Z25152
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
Contains 1 vial of Zenon Biotin-XX mouse IgG2a labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 250 μL).
Generera snabbt antikroppsisotypkonjugat för immuncytokemi (ICC), immunhistokemi (IHC), flödescytometri och cellavbildning med Invitrogen Zenon™ mus IgG 2a märkningssatser.
Generera snabbt antikroppsisotypkonjugat för immuncytokemi (ICC), immunhistokemi (IHC), flödescytometri och cellavbildning med Invitrogen Zenon™ mus IgG 2a märkningssatser. Förenkla dina laboratorieapplikationer och minska korsreaktiviteten av antikroppar samtidigt som du uppnår effektiv märkning av antikroppar i serum, ascitesvätska eller hybridomsuspensioner.
Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled IgG2a primary antibodies with the Zenon™ mouse IgG2a labeling kits. These kits utilize Alexa Fluor fluorophores, biotin, Pacific Blue, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG2a primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.
Zenon labeling technology greatly simplifies the use of multiple mouse-derived antibodies in the same staining protocol. Zenon tricolor labeling kits contain sufficient materials for 10 labeling reactions of each of three different fluorescent colors.
Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, HRP, alkaline phosphatase
• Multiplex with other mouse monoclonal antibodies simultaneously
• Can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.
Advantages of using Zenon antibody labeling kits:
Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.
Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.
Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.
Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.
Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.
Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 50 Reactions
Innehåll och lagring Contains 1 vial of Zenon Biotin-XX mouse IgG2a labeling reagent (250 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 250 μL).
Store in refrigerator (2–8°C) and protect from light.
Märkning Skala < 1–20 μg
Produktlinje Zenon
Etikett eller färgämne Biotin-XX
Färg Colorless
Märkningsmål IgG2a
Art Mouse
Etiketttyp Biotin & Other Haptens