missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17187-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.Specifications
ZFP64 | |
Polyclonal | |
Unconjugated | |
ZFP64 | |
Zfp 64, ZFP64, Zinc finger protein 338, ZNF338 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22722, 311661, 55734 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99KE8, Q9NPA5 | |
Zfp64 | |
ZFP64 Fusion Protein Ag10971 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |