missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ZHX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570373
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This target displays homology in the following species: Guinea Pig: 86%; Human: 100%; Rabbit: 92%; Rat: 86%.
This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor.
Specifikationer
| ZHX3 | |
| Polyclonal | |
| Unconjugated | |
| ZHX3 | |
| 1810059C13Rik; 4932418O04Rik; 9530010N21Rik; KIAA0395; mKIAA0395; Tix1; triple homeobox 1; triple homeobox protein 1; Zhx3; Zinc finger and homeodomain protein 3; zinc fingers and homeoboxes 3; zinc fingers and homeoboxes protein 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23051 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4I2 | |
| ZHX3 | |
| synthetic peptide directed towards the middle region of human ZHX3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering