Läs mer
Zinc 2-Mercaptobenzothiazole, TRC
Produktkod. 30133297
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133297 5 g
30138337 50 g
30003772 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133297 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ7015685G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 155-04-4
Molekylformel C14H8N2S4Zn
Molekylvikt (g/mol) 397.85
IUPAC-namn zinc;1,3-benzothiazole-2-thiolate
LEDER [Zn+2].[S-]c1nc2ccccc2s1.[S-]c3nc4ccccc4s3
Specifikationer
Kemiskt namn eller material Zinc 2-Mercaptobenzothiazole
Kvantitet 5 g
InChI formel InChI=1S/2C7H5NS2.Zn/c2*9-7-8-5-3-1-2-4-6(5)10-7;/h2*1-4H,(H,8,9);/q;;+2/p-2
Formel vikt 395.8862