missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc for AAS 10g/L HCl matrix, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
Packaging:
Bottle
Specifications
Concentration | 10g/L ±0.02g/L |
CAS | 7697-37-2 |
Packaging | Bottle |
Quantity | 100 mL |
Type | Single-element Concentrated Standard Solution |
Molecular Formula | HNO3 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
SMILES | O[N+]([O-])=O |
IUPAC Name | nitric acid |
Show More |
Safety and Handling
Product Identifier
- Single-element Standard Solution for AAS. Zinc (Zn) 10000mg/l in HCl 5%
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'