Läs mer
Zinc Bis(2-Ethylhexyl) Phosphorodithioate, TRC
Produktkod. 30452561
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30452561 50 mg
1 options
Produktkod. 30452561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ43846050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zinc Bis(2-Ethylhexyl) Phosphorodithioate
CAS 4259-15-8
Molekylformel C32H68O4P2S4Zn
Kvantitet 50 mg
Synonym Acorox 880, Additin RC 1529, Bis(2-ethylhexyl) phosphorodithioate Zinc Salt, DDP-Zn-O, Elco 108, Multadit OB, RC 3180, RC 31800, ZPTP 3, Zinc O,O-bis(2-ethylhexyl) phosphorodithioate, Zinc Bis(2-ethylhexyl) phosphorodithioateZinc di(2-Ethylhexyl) Dithiophosphate, Zinc o,o'-di-2-Ethylhexyldithiophosphate
InChI formel InChI=1S/2C16H35O2PS2.Zn/c2*1-5-9-11-15(7-3)13-17-19(20,21)18-14-16(8-4)12-10-6-2;/h2*15-16H,5-14H2,1-4H3,(H,20,21);/q;;+2/p-2
LEDER [Zn+2].CCCCC(CC)COP(=S)([S-])OCC(CC)CCCC.CCCCC(CC)COP(=S)([S-])OCC(CC)CCCC
IUPAC-namn zinc;bis(2-ethylhexoxy)-sulfanylidene-sulfido-λ^{5}-phosphane
Molekylvikt (g/mol) 772.47
Formel vikt 770.2767