missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 1.9M in 2-MeTHF, Thermo Scientific Chemicals

Catalog Number 15400708
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7646-85-7
Quantity 100 mL
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
UN Number UN2924
Merck Index 14,10132
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Molecular Weight (g/mol) 136.28
PubChem CID 5727
ChEBI CHEBI:49976
Formula Weight 136.29
Concentration or Composition (by Analyte or Components) 1.9M in 2-MeTHF
Sensitivity Air and moisture sensitive
Density 1.07
Chemical Name or Material Zinc chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'