missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride ACS AR Granular, Macron Fine Chemicals™

macron

Brand:  Macron Fine Chemicals 8780-04

Additional Details : CAS Number : 7646-85-7 Weight : 0.60000kg Transport : UN number : 2331 Chem class : 8 Pack group : III

Packaging POLYSTORMOR
Quantity 500g

 View more versions of this product

Product Code. 15568764

  • 1949.00 SEK / 500g


Estimated Shipment
27-01-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

7646-85-7
136.28
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
CHEBI:49976
Cl[Zn]Cl
Cl2Zn
JIAARYAFYJHUJI-UHFFFAOYSA-L
5727
dichlorozinc

Specifications

Zinc Chloride
Cl2Zn
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.3
ACS
500g
7646-85-7
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Cl[Zn]Cl
136.28
CHEBI:49976
Granular
POLYSTORMOR
Videos
SDS
Documents