Läs mer
Zinc Cysteinate Hydrochloride, TRC
Produktkod. 30453967
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30453967 100 mg
1 options
Produktkod. 30453967 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ439600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Zinc Cysteinate Hydrochloride
CAS 549521-94-0
Molekylformel C3 H5 N O2 S . H Cl . Zn
Kvantitet 100 mg
Synonym [L-Cysteinato(2-)-κO,κS]zinc Hydrochloride
InChI formel InChI=1S/C3H7NO2S.ClH.Zn/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;/q;;+2/p-2/t2-;;/m0../s1
LEDER Cl.[Zn+2].N[C@@H](C[S-])C(=O)[O-]
IUPAC-namn zinc;(2R)-2-amino-3-sulfidopropanoate;hydrochloride
Molekylvikt (g/mol) 221.012
Formel vikt 218.91