Läs mer
Zinc Difluoromethanesulfinate (Contains up to 1 equiv. of Zinc Chloride), TRC
Produktkod. 30452865
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452865 50 mg
30455228 100 mg
30453207 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30452865 Leverantör Toronto Research Chemicals Leverantörsnummer TRCZ70169350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1355729-38-2
Molekylformel C2H2F4O4S2Zn
Molekylvikt (g/mol) 295.54
Synonym Baran Reagent
IUPAC-namn zinc;difluoromethanesulfinate
LEDER [Zn+2].[O-]S(=O)C(F)F.[O-]S(=O)C(F)F
Specifikationer
Kemiskt namn eller material Zinc Difluoromethanesulfinate
Kvantitet 50 mg
InChI formel InChI=1S/2CH2F2O2S.Zn/c2*2-1(3)6(4)5;/h2*1H,(H,4,5);/q;;+2/p-2
Formel vikt 293.8622