Läs mer
Zinc Dihydrogen Phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Zinc Dihydrogen Phosphate |
| CAS | 13598-37-3 |
| Molekylformel | ZnH4P2O8 |
| Kvantitet | 5 g |
| Synonym | Zinc phosphate (Zn(H2PO4)2) (6CI','7CI), Budit T 21, Monobasic zinc phosphate, Z 21-82, Zinc bis(dihydrogen phosphate), Zinc diacid phosphate, Zinc dihydrogen phosphate, Zinc dihydrogen phosphate (Zn(H2PO4)2), Zinc dihydrophosphate, Zinc hydrogen phosphate (Zn(H2PO4)2), Zinc phosphate (1:2), Zinc primary phosphate |
| InChI formel | InChI=1S/2H3O4P.Zn/c2*1-5(2,3)4;/h2*(H3,1,2,3,4);/q;;+2/p-2 |
| LEDER | OP(O)([O-])=O.OP(O)([O-])=O.[Zn+2] |
| IUPAC-namn | zinc(II) dihydrogen phosphate |
| Molekylvikt (g/mol) | 259.35 |
| Formel vikt | 257.87 |