missing translation for 'onlineSavingsMsg'
Learn More

Zinc hexafluoro-2,4-pentanedionate dihydrate, Thermo Scientific Chemicals

Catalog Number p-7050829
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16743-33-2
C10H6F12O6Zn
515.51
MFCD00150577
HHDNHEKLDWSJAJ-LJDKTGGESA-L
zinc hexafluoroacetylacetonate hydrate, zinc hexafluoroacetylacetonate dihydrate, zinc hexafluoro 2,4-pentanedionate, zinc hexafluoro-2,4-pentanedionate dihydrate, hexafluoroacetylacetone zinc derivative dihydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy zincio oxy pent-3-en-2-one dihydrate
72376390
O.O.[Zn++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
16743-33-2
C10H6F12O6Zn
515.51
MFCD00150577
HHDNHEKLDWSJAJ-LJDKTGGESA-L
zinc hexafluoroacetylacetonate hydrate, zinc hexafluoroacetylacetonate dihydrate, zinc hexafluoro 2,4-pentanedionate, zinc hexafluoro-2,4-pentanedionate dihydrate, hexafluoroacetylacetone zinc derivative dihydrate, 3z-1,1,1,5,5,5-hexafluoro-4-2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy zincio oxy pent-3-en-2-one dihydrate
72376390
O.O.[Zn++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 158°C
Physical Form Powder
Quantity 1 g
IUPAC Name zinc(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) dihydrate
Formula Weight 515.49 (479.47 Anhydrous)
Chemical Name or Material Zinc hexafluoro-2,4-pentanedionate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'